EMAIL THIS PAGE TO A FRIEND
8.01961 EMD Millipore

1-Bromoundecane

1-Bromoundecane for synthesis. CAS 693-67-4, chemical formula CH₃(CH₂)10Br., for synthesis

Synonym: Undecyl bromide

  • CAS Number 693-67-4

  • Linear Formula C₁₁H₂₃Br

  • Molecular Weight 235.2 g/mol

  •  EC Number 211-757-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8019610025)
bp   137 - 138 °C (24 hPa)
density   1.05 g/cm3 (20 °C)
mp   -9 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.052 - 1.058
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?