• USA Home
  • 8.01975 - 5-Bromo-2-hydroxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.01975 EMD Millipore

5-Bromo-2-hydroxybenzoic acid

5-Bromo-2-hydroxybenzoic acid for synthesis. CAS 89-55-4, molar mass 217.02 g/mol., for synthesis

Synonym: 5-Bromsalicylic acid

  • CAS Number 89-55-4

  • Linear Formula C₇H₅BrO₃

  • Molecular Weight 217.02 g/mol

  •  EC Number 201-917-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8019750100)
mp   166 - 168 °C
shipped in   ambient
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (upper value): ≥ 163 °C
  Melting range (lower value): ≥ 167 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 29 00
RTECS 
VO1850000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?