EMAIL THIS PAGE TO A FRIEND
8.02226 EMD Millipore

Benzyl mercaptan

Benzyl mercaptan for synthesis. CAS No. 100-53-8, EC Number 202-862-5., for synthesis

Synonym: Thiobenzyl alcohol, α-Toluenethiol

  • CAS Number 100-53-8

  • Linear Formula C₇H₈S

  • Molecular Weight 124.2 g/mol

  •  EC Number 202-862-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8022260250)
bp   193 - 195 °C (1013 hPa)
expl. lim.     1 %(V)
vapor pressure   0.5 hPa (20 °C)
density   1.055 g/cm3 (20 °C)
flash point   70 °C
autoignition temp.   270 °C
mp   -29.2 °C
LD 50 oral   LD50 Rat 493 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.054 - 1.056
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P260: Do not breathe vapour.
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 22-50/53
Harmful if swallowed.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 23-24/25-61
Do not breathe vapour.Avoid contact with skin and eyes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?