EMAIL THIS PAGE TO A FRIEND
8.02406 EMD Millipore

Isoquinoline

Isoquinoline for synthesis. CAS 119-65-3, pH 7.5 (5 g/l, H₂O, 20 °C). MSDS and product info below., for synthesis

  • CAS Number 119-65-3

  • Linear Formula C₉H₇N

  • Molecular Weight 129.16 g/mol

  •  EC Number 204-341-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   243 °C (1013 hPa)
vapor pressure   0.05 hPa (20 °C)
solubility   5 g/l
density   1.09 g/cm3 (20 °C)
flash point   107 °C
mp   21 - 26 °C
pH   7.5 (5 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 615 mg/kg
LD 50 dermal   LD50 Rat 180 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Melting range (lower value): ≥ 18 °C
  Melting range (upper value): ≤ 21 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 49 90
Storage class 
6.1B Non combustible substances, toxic
WGK 
WGK 2 water endangering
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
toxic
R Phrase 
R 22-24
Harmful if swallowed.Toxic in contact with skin.
S Phrase 
S 24/25-36/37-45
Avoid contact with skin and eyes.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?