• USA Home
  • 8.02452 - 3-Chlorobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.02452 EMD Millipore

3-Chlorobenzaldehyde

3-Chlorobenzaldehyde for synthesis. CAS No. 587-04-2, EC Number 209-596-9., for synthesis

  • CAS Number 587-04-2

  • Linear Formula C₇H₅ClO

  • Molecular Weight 140.57 g/mol

  •  EC Number 209-596-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   214 °C
density   1.24 g/cm3 (20 °C)
flash point   88 °C
mp   17 - 18 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.234 - 1.240
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2913 00 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?