EMAIL THIS PAGE TO A FRIEND
8.02460 EMD Millipore

Cycloheptanol

Cycloheptanol for synthesis. CAS 502-41-0, molar mass 114.18 g/mol., for synthesis

  • CAS Number 502-41-0

  • Linear Formula C₇H₁₄O

  • Molecular Weight 114.18 g/mol

  •  EC Number 207-936-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   185 °C (1013 hPa)
density   0.96 g/cm3 (20 °C)
flash point   64 °C
mp   2 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.958 - 0.960
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 19 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?