EMAIL THIS PAGE TO A FRIEND
8.02462 EMD Millipore

Cycloheptanone

Cycloheptanone for synthesis. CAS 502-42-1, molar mass 112.17 g/mol., for synthesis

Synonym: Suberone

  • CAS Number 502-42-1

  • Linear Formula C₇H₁₂O

  • Molecular Weight 112.17 g/mol

  •  EC Number 207-937-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8024620050)
density   0.95 g/cm3 (20 °C)
flash point   55 °C
mp   -21 °C
LD 50 oral   LD50 Rat 500 - 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.949 - 0.953
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable, harmful
R Phrase 
R 10-22
Flammable.Harmful if swallowed.
S Phrase 
S 23-24/25
Do not breathe vapour.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?