EMAIL THIS PAGE TO A FRIEND
8.02774 EMD Millipore

Bromocyclopentane

Bromocyclopentane for synthesis. CAS 137-43-9, molar mass 149.03 g/mol., for synthesis

Synonym: Cyclopentyl bromide

  • CAS Number 137-43-9

  • Linear Formula C₅H₉Br

  • Molecular Weight 149.03 g/mol

  •  EC Number 205-294-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8027740100)
bp   137 - 139 °C (1013 hPa)
density   1.39 g/cm3 (20 °C)
flash point   35 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.390 - 1.393
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 89 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P260: Do not breathe vapour.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 23-24/25
Do not breathe vapour.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?