• USA Home
  • 8.02794 - 3-Chloropropionitrile

EMAIL THIS PAGE TO A FRIEND
8.02794 EMD Millipore

3-Chloropropionitrile

3-Chloropropionitrile for synthesis. CAS No. 542-76-7, EC Number 208-827-0., for synthesis

Synonym: 3-Chloropropionic acid nitrile

  • CAS Number 542-76-7

  • Linear Formula C₃H₄ClN

  • Molecular Weight 89.52 g/mol

  •  EC Number 208-827-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8027940250)
bp   174 - 176 °C (1013 hPa)
vapor pressure   8 hPa (50 °C)
solubility   45 g/l
density   1.14 g/cm3 (20 °C)
flash point   76 °C
mp   -52 °C
LD 50 oral   LD50 Rat 10 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.143 - 1.146
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H300: Fatal if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Very toxic  Very toxic
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, II
Categories of danger 
very toxic, irritant
R Phrase 
R 28-36/38
Very toxic if swallowed.Irritating to eyes and skin.
S Phrase 
S 26-28-37-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.After contact with skin, wash immediately with plenty of water.Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?