• USA Home
  • 8.02812 - 5-Chlorovaleric acid

EMAIL THIS PAGE TO A FRIEND
8.02812 EMD Millipore

5-Chlorovaleric acid

5-Chlorovaleric acid for synthesis. CAS No. 1119-46-6, EC Number 214-279-3., for synthesis

Synonym: 5-Chloropentanoic

  • CAS Number 1119-46-6

  • Linear Formula C₅H₉ClO₂

  • Molecular Weight 136.57 g/mol

  •  EC Number 214-279-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8028120025)
bp   138 - 140 °C (13 hPa)
density   1.17 g/cm3 (20 °C)
flash point   >110 °C
mp   17 - 19 °C
pH   2.5 (100 g/l, H₂O, 20 °C) (as an emulsion)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.168 - 1.170
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?