• USA Home
  • 8.02823 - Cyclohexanecarbonitrile

EMAIL THIS PAGE TO A FRIEND
8.02823 EMD Millipore

Cyclohexanecarbonitrile

Cyclohexanecarbonitrile for synthesis. CAS 766-05-2, chemical formula C₆H₁₁CN., for synthesis

Synonym: Hexahydrobenzonitrile, Cyclohexyl cyanide

  • CAS Number 766-05-2

  • Linear Formula C₇H₁₁N

  • Molecular Weight 109.17 g/mol

  •  EC Number 212-157-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8028230025)
density   0.92 g/cm3 (20 °C)
flash point   65 °C
mp   12 °C
LD 50 oral   LD50 Rat 80 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 94.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.913 - 0.915
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H301: Toxic if swallowed.
Precautionary Statement(s) 
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, III
Categories of danger 
toxic
R Phrase 
R 25
Toxic if swallowed.
S Phrase 
S 37-45
Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?