EMAIL THIS PAGE TO A FRIEND
8.02866 EMD Millipore

Cyanoacetamide

Cyanoacetamide for synthesis. CAS 107-91-5, chemical formula NCCH₂CONH₂., for synthesis

  • CAS Number 107-91-5

  • Linear Formula C₃H₄N₂O

  • Molecular Weight 84.07 g/mol

  •  EC Number 203-531-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Plastic bottle (8028660250)
solubility   150 g/l
flash point   215 °C
mp   117 - 120 °C
LD 50 oral   LD50 Rat 7230 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 98.0 %
  Melting range (lower value): ≥ 118 °C
  Melting range (upper value): ≤ 121 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?