EMAIL THIS PAGE TO A FRIEND
8.02874 EMD Millipore

Methyl carbamate

Methyl carbamate for synthesis. CAS 598-55-0, EC Number 209-939-2, chemical formula NH₂COOCH₃., for synthesis

Synonym: Carbamic acid methyl ester

  • CAS Number 598-55-0

  • Linear Formula C₂H₅NO₂

  • Molecular Weight 75.07 g/mol

  •  EC Number 209-939-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8028740005)
bp   176 - 178 °C
solubility   700 g/l
flash point   >100 °C
mp   54 - 56 °C
pH   6 - 8 (50 g/l, H₂O, 20 °C)
bulk density   500 kg/m3
LD 50 dermal   LD50 Rabbit > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 98.0 %
  Melting range (lower value): ≥ 54 °C
  Melting range (upper value): ≤ 57 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?