EMAIL THIS PAGE TO A FRIEND
8.02884 EMD Millipore

Cyclohexanethiol

Cyclohexanethiol for synthesis. CAS 1569-69-3, chemical formula C₆H₁₁SH., for synthesis

Synonym: Cyclohexyl mercaptan

  • CAS Number 1569-69-3

  • Linear Formula C₆H₁₂S

  • Molecular Weight 116.23 g/mol

  •  EC Number 216-378-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 ml in Glass bottle (8028840500)
bp   158 - 160 °C (1013 hPa)
expl. lim.     1.1 %(V)
vapor pressure   5 hPa (20 °C)
solubility   0.05 g/l slightly soluble
density   0.95 g/cm3 (20 °C)
flash point   43 °C
mp   <-20 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.946 - 0.948
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H331: Toxic if inhaled.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat.
P273: Avoid release to the environment.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3054 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3054 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3054 , 3, III
Categories of danger 
flammable, irritant, dangerous for the environment
R Phrase 
R 10-20/22-38-43-51/53
Flammable.Harmful by inhalation and if swallowed.Irritating to skin.May cause sensitisation by skin contact.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 23-24-37-61
Do not breathe vapour.Avoid contact with skin.Wear suitable gloves.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?