• USA Home
  • 8.02927 - (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

EMAIL THIS PAGE TO A FRIEND
8.02927 EMD Millipore

(2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

(2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate for resolution of racemates for synthesis. CAS 62708-56-9, chemical formula C₆H₅COOCH(COOH)CH(COOH)OCOC₆H₅., for resolution of racemates for synthesis

  • CAS Number 62708-56-9

  • Linear Formula C₁₈H₁₄O₈*H₂O

  • Molecular Weight 376.32 g/mol

  •  EC Number 220-374-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8029270100)
mp   87 - 90 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 87 °C
  Melting range (upper value): ≤ 90 °C
  Spec. rotation [α²0/D (c=5 in methanol): -119.0 - -116.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 13 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?