EMAIL THIS PAGE TO A FRIEND
8.02965 EMD Millipore

Fluorobenzene

Fluorobenzene for synthesis. CAS 462-06-6, molar mass 96.1 g/mol., for synthesis

  • CAS Number 462-06-6

  • Linear Formula C₆H₅F

  • Molecular Weight 96.1 g/mol

  •  EC Number 207-321-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8029650005)
expl. lim.     1.3 - 8.9 %(V)
vapor pressure   83 hPa (20 °C)
solubility   1.54 g/l
density   1.03 g/cm3 (20 °C)
flash point   -15 °C
autoignition temp.   630 °C
mp   -41 °C
LD 50 oral   LD50 Rat 4399 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.024 - 1.026
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P233: Keep container tightly closed.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 2387 , 3, II
Declaration (transport by air) IATA-DGR 
UN 2387 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 2387 , 3, II, Segregation Group: 10 (Liquid halogenated hydrocarbons)
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16-29-33
Keep away from sources of ignition - No smoking.Do not empty into drains.Take precautionary measures against static discharges.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?