• USA Home
  • 8.03030 - 1,3-Dichloro-2-propanol

EMAIL THIS PAGE TO A FRIEND
8.03030 EMD Millipore

1,3-Dichloro-2-propanol

1,3-Dichloro-2-propanol for synthesis. CAS 96-23-1, molar mass 128.98 g/mol., for synthesis

Synonym: Glycerol α,γ-dichlorohydrin, α-Dichlorohydrin

  • CAS Number 96-23-1

  • Linear Formula C₃H₆Cl₂O

  • Molecular Weight 128.98 g/mol

  •  EC Number 202-491-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8030300005)
bp   175 °C (1013 hPa)
vapor pressure   0.72 hPa (20 °C)
solubility   110 g/l
density   1.36 g/cm3 (20 °C)
flash point   85 °C
mp   -4 °C
LD 50 oral   LD50 Rat 110 mg/kg
LD 50 dermal   LD50 Rabbit 800 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.363 - 1.365
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 59 98
Hazard Statement(s) 
H350: May cause cancer.
H301: Toxic if swallowed.
H312: Harmful in contact with skin.
Precautionary Statement(s) 
P201: Obtain special instructions before use.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
UB1400000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2750 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2750 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2750 , 6.1, II
Categories of danger 
carcinogenic, toxic
R Phrase 
R 45-21-25
May cause cancer.Also harmful in contact with skin.Also toxic if swallowed.
S Phrase 
S 53-45
Avoid exposure - obtain special instructions before use.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?