• USA Home
  • 8.03031 - 2,3-Dichloro-1-propanol

EMAIL THIS PAGE TO A FRIEND
8.03031 EMD Millipore

2,3-Dichloro-1-propanol

2,3-Dichloro-1-propanol for synthesis. CAS 616-23-9, chemical formula ClCH₂CHClCH₂OH., for synthesis

Synonym: Glycerol-α,ß-dichlorohydrin

  • CAS Number 616-23-9

  • Linear Formula C₃H₆Cl₂O

  • Molecular Weight 128.98 g/mol

  •  EC Number 210-470-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8030310010)
vapor pressure   0.245 hPa (25 °C)
solubility   1 - 10 g/l
density   1.36 g/cm3 (20 °C)
flash point   93 °C
LD 50 oral   LD50 Rat 90 mg/kg
LD 50 dermal   LD50 Rabbit 200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.359 - 1.364
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 59 98
Hazard Statement(s) 
H301: Toxic if swallowed.
H310: Fatal in contact with skin.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe vapour.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
toxic, irritant
R Phrase 
R 24/25-36
Toxic in contact with skin and if swallowed.Irritating to eyes.
S Phrase 
S 23-26-36/37-45
Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?