• USA Home
  • 8.03036 - Bis(cyclopentadienyl)titanium dichloride

EMAIL THIS PAGE TO A FRIEND
8.03036 EMD Millipore

Bis(cyclopentadienyl)titanium dichloride

Bis(cyclopentadienyl)titanium dichloride for synthesis. CAS No. 1271-19-8, EC Number 215-035-9., for synthesis

Synonym: Dichlorodicyclopentadienyltitanium, Titanocene dichloride

  • CAS Number 1271-19-8

  • Linear Formula C₁₀H₁₀Cl₂Ti

  • Molecular Weight 248.96 g/mol

  •  EC Number 215-035-9

Purchase

Properties

packaging   5 g in Glass bottle (8030360005)
mp   289 - 291 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex Cl): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?