• USA Home
  • 8.03066 - N,N-Dimethylformamide diethyl acetal

EMAIL THIS PAGE TO A FRIEND
8.03066 EMD Millipore

N,N-Dimethylformamide diethyl acetal

N,N-Dimethylformamide diethyl acetal for synthesis. CAS 1188-33-6, EC Number 214-707-9, chemical formula (C₂H₅O)₂CHN(CH₃)₂., for synthesis

Synonym: N,N-Dimethyldiethoxymethylamine

  • CAS Number 1188-33-6

  • Linear Formula C₇H₁₇NO₂

  • Molecular Weight 147.21 g/mol

  •  EC Number 214-707-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8030660025)
density   0.86 g/cm3 (20 °C)
flash point   23 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 90.0 %
  Density (d 20 °C/ 4 °C): 0.859 - 0.866
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 50 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?