• USA Home
  • 8.03461 - Ethyl 1,3-dioxoisoindoline 2-carboxylate

EMAIL THIS PAGE TO A FRIEND
8.03461 EMD Millipore

Ethyl 1,3-dioxoisoindoline 2-carboxylate

Ethyl 1,3-dioxoisoindoline 2-carboxylate for synthesis. CAS 22509-74-6, molar mass 219.19 g/mol., for synthesis

Synonym: Phthalimide-N-carboxylic acid ethyl ester, 1,3-Dioxo-2-isoindolinecarboxylic acid ethyl ester, N-Ethoxycarbonylphthalimide

  • CAS Number 22509-74-6

  • Linear Formula C₁₁H₉NO₄

  • Molecular Weight 219.19 g/mol

  •  EC Number 245-048-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8034610025)
mp   85 - 90 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 85 °C
  Melting range (upper value): ≤ 90 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2925 19 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?