• USA Home
  • 8.04528 - N-Hydroxyphthalimide

EMAIL THIS PAGE TO A FRIEND
8.04528 EMD Millipore

N-Hydroxyphthalimide

N-Hydroxyphthalimide for synthesis. CAS 524-38-9, molar mass 163.13 g/mol., for synthesis

  • CAS Number 524-38-9

  • Linear Formula C₈H₅NO₃

  • Molecular Weight 163.13 g/mol

  •  EC Number 208-358-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8045280100)
density   1.64 g/cm3
mp   233 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 230 °C
  Melting range (upper value): ≤ 235 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?