EMAIL THIS PAGE TO A FRIEND
8.04533 EMD Millipore

3-Hexanone

3-Hexanone for synthesis. CAS 589-38-8, molar mass 100.16 g/mol., for synthesis

Synonym: Ethyl propyl ketone

  • CAS Number 589-38-8

  • Linear Formula C₆H₁₂O

  • Molecular Weight 100.16 g/mol

  •  EC Number 209-645-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8045330050)
bp   119 - 121 °C (1011 hPa)
expl. lim.     1 - 8 %(V)
vapor pressure   18 hPa (20 °C)
solubility   14.7 g/l
density   0.81 g/cm3 (20 °C)
flash point   14 °C
mp   -55 °C
LD 50 oral   LD50 Rat 2738 mg/kg
LD 50 dermal   LD50 Rabbit 2583 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.814 - 0.816
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 19 90
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
P240: Ground/bond container and receiving equipment.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
RTECS 
MP1575000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1224 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1224 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1224 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16
Keep away from sources of ignition - No smoking.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?