• USA Home
  • 8.04535 - 3-Hydroxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.04535 EMD Millipore

3-Hydroxybenzaldehyde

3-Hydroxybenzaldehyde for synthesis. CAS No. 100-83-4, EC Number 202-892-9., for synthesis

  • CAS Number 100-83-4

  • Linear Formula C₇H₆O₂

  • Molecular Weight 122.12 g/mol

  •  EC Number 202-892-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8045350100)
bp   191 °C (67 hPa)
solubility   7.2 g/l
autoignition temp.   >400 °C
mp   99 - 102 °C
pH   3 - 4 (H₂O, 20 °C) (saturated solution)
bulk density   350 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 100 °C
  Melting range (upper value): ≤ 104 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 49 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe dust.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?