• USA Home
  • 8.04543 - 3-Phenylpropionic acid

EMAIL THIS PAGE TO A FRIEND
8.04543 EMD Millipore

3-Phenylpropionic acid

3-Phenylpropionic acid for synthesis. CAS No. 501-52-0, EC Number 207-924-5., for synthesis

Synonym: Hydrocinnamic acid

  • CAS Number 501-52-0

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.17 g/mol

  •  EC Number 207-924-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8045430100)
bp   279 - 281 °C (1013 hPa)
solubility   5.9 g/l soluble
density   1.071 g/cm3
flash point   110 °C
autoignition temp.   505 °C
mp   47 - 49 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 47 °C
  Melting range (upper value): ≤ 49 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe dust.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?