EMAIL THIS PAGE TO A FRIEND
8.04554 EMD Millipore

1-Bromoheptane

1-Bromoheptane for synthesis. CAS 629-04-9, chemical formula CH₃(CH₂)₆Br., for synthesis

Synonym: Heptyl bromide

  • CAS Number 629-04-9

  • Linear Formula C₇H₁₅Br

  • Molecular Weight 179.09 g/mol

  •  EC Number 211-068-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8045540250)
solubility   0.0066 g/l
density   1.14 g/cm3 (20 °C)
flash point   60 °C
mp   -58 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.137 - 1.140
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
R Phrase 
R 10
Flammable.
S Phrase 
S 15
Keep away from heat.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?