• USA Home
  • 8.04605 - Hydrazinium monobromide

EMAIL THIS PAGE TO A FRIEND
8.04605 EMD Millipore

Hydrazinium monobromide

Hydrazinium monobromide for synthesis. CAS 13775-80-9, EC Number 237-412-7, chemical formula NH₂NH₃Br., for synthesis

Synonym: Hydrazine monohydrobromide

  • CAS Number 13775-80-9

  • Linear Formula H₅BrN₂

  • Molecular Weight 112.96 g/mol

  •  EC Number 237-412-7

Purchase

Properties

packaging   250 g in Plastic bottle (8046050250)
solubility   3260 g/l soluble
mp   85 °C
pH   4 - 5 (100 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (iodometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2825 10 00
Hazard Statement(s) 
H350: May cause cancer.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
H317: May cause an allergic skin reaction.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P201: Obtain special instructions before use.
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3288 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3288 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3288 , 6.1, III
Categories of danger 
carcinogenic, toxic, sensitizing, dangerous for the environment
R Phrase 
R 45-23/24/25-43-50/53
May cause cancer.Also toxic by inhalation, in contact with skin and if swallowed.May cause sensitisation by skin contact.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 53-45-60-61
Avoid exposure - obtain special instructions before use.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).This material and its container must be disposed of as hazardous waste.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?