• USA Home
  • 8.04613 - Hydroquinone dimethyl ether

EMAIL THIS PAGE TO A FRIEND
8.04613 EMD Millipore

Hydroquinone dimethyl ether

Hydroquinone dimethyl ether for synthesis. CAS 150-78-7, EC Number 205-771-9, chemical formula C₆H₄-1,4-(OCH₃)₂., for synthesis

Synonym: 1,4-Dimethoxybenzene

  • CAS Number 150-78-7

  • Linear Formula C₈H₁₀O₂

  • Molecular Weight 138.17 g/mol

  •  EC Number 205-771-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8046130005)
bp   213 °C (1013 hPa)
expl. lim.     1.2 - 5.6 %(V)
vapor pressure   <0.1 hPa (20 °C)
solubility   0.80 g/l
flash point   94 °C
autoignition temp.   438 °C
mp   >53 - 56 °C
pH   7 (H₂O, 20 °C) (saturated solution)
bulk density   600 kg/m3
LD 50 oral   LD50 Rat 3600 mg/kg
LD 50 dermal   LD50 Rat > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 53 °C
  Melting range (upper value): ≤ 56 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
RTECS 
CZ6650000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?