EMAIL THIS PAGE TO A FRIEND
8.04732 EMD Millipore

Indoline

Indoline for synthesis. CAS 496-15-1, molar mass 119.17 g/mol., for synthesis

Synonym: 2,3-Dihydroindole

  • CAS Number 496-15-1

  • Linear Formula C₈H₉N

  • Molecular Weight 119.17 g/mol

  •  EC Number 207-816-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8047320025)
bp   220 - 221 °C (1013 hPa)
solubility   5 g/l
density   1.06 g/cm3 (20 °C)
flash point   103 °C
autoignition temp.   460 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.062 - 1.064
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
RTECS 
NL6906300
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?