• USA Home
  • 8.04781 - Iodosobenzene diacetate

EMAIL THIS PAGE TO A FRIEND
8.04781 EMD Millipore

Iodosobenzene diacetate

Iodosobenzene diacetate for synthesis. CAS 3240-34-4, EC Number 221-808-1, chemical formula C₆H₅I(OCOCH₃)₂., for synthesis

Synonym: Diacetoxyiodosobenzene, Phenyliodine diacetate

  • CAS Number 3240-34-4

  • Linear Formula C₁₀H₁₁IO₄

  • Molecular Weight 322.09 g/mol

  •  EC Number 221-808-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8047810010)
mp   161 - 164 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (iodometric): ≥ 99.0 %
  Melting range (lower value): ≥ 161 °C
  Melting range (upper value): ≤ 164 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?