EMAIL THIS PAGE TO A FRIEND
8.04787 EMD Millipore

2-Iodobutane

2-Iodobutane (stabilised with sodium carbonate) for synthesis. CAS 513-48-4, chemical formula CH₃CH₂CH(I)CH₃., (stabilised with sodium carbonate) for synthesis

Synonym: sec-Butyl iodide

  • CAS Number 513-48-4

  • Linear Formula C₄H₉I

  • Molecular Weight 184.02 g/mol

  •  EC Number 208-163-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8047870050)
bp   119 - 120 °C (1013 hPa)
density   1.59 g/cm3 (20 °C)
flash point   21 °C
mp   -104 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (20/4 °C): 1.593 - 1.596
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 80
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P260: Do not breathe vapour.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 2390 , 3, II
Declaration (transport by air) IATA-DGR 
UN 2390 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 2390 , 3, II, Segregation Group: 10 (Liquid halogenated hydrocarbons)
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
S Phrase 
S 23
Do not breathe vapour.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?