• USA Home
  • 8.04791 - Dimethyl isophthalate

EMAIL THIS PAGE TO A FRIEND
8.04791 EMD Millipore

Dimethyl isophthalate

Dimethyl isophthalate for synthesis. CAS 1459-93-4, molar mass 194.18 g/mol., for synthesis

Synonym: Isophthalic acid dimethyl ester

  • CAS Number 1459-93-4

  • Linear Formula C₁₀H₁₀O₄

  • Molecular Weight 194.18 g/mol

  •  EC Number 215-951-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8047910100)
bp   282 °C (1013 hPa)
vapor pressure   0.012 hPa (25 °C)
solubility   0.29 g/l insoluble
density   1.194 g/cm3 (20 °C)
flash point   138 °C
LD 50 oral   LD50 Rat 4390 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 65 °C
  Melting range (upper value): ≤ 68 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
RTECS 
NT2540000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?