• USA Home
  • 8.04802 - Indole-3-acetonitrile

EMAIL THIS PAGE TO A FRIEND
8.04802 EMD Millipore

Indole-3-acetonitrile

Indole-3-acetonitrile for synthesis. CAS 771-51-7, EC Number 212-232-1, molar mass 156.19 g/mol., for synthesis

Synonym: 3-Indolylacetonitrile

  • CAS Number 771-51-7

  • Linear Formula C₁₀H₈N₂

  • Molecular Weight 156.19 g/mol

  •  EC Number 212-232-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8048020010)
bp   156 - 160 °C (0.27 hPa)
flash point   206 °C
autoignition temp.   540 °C
mp   33 - 36 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 33 °C
  Melting range (upper value): ≤ 36 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?