• USA Home
  • 8.04804 - Ethyl 3-indoleacetate

EMAIL THIS PAGE TO A FRIEND
8.04804 EMD Millipore

Ethyl 3-indoleacetate

Ethyl 3-indoleacetate for synthesis. CAS 778-82-5, molar mass 203.24 g/mol., for synthesis

Synonym: 3-Indole acetic acid ethyl ester

  • CAS Number 778-82-5

  • Linear Formula C₁₂H₁₃NO₂

  • Molecular Weight 203.24 g/mol

  •  EC Number 212-296-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8048040005)
bp   164 - 166 °C (1.33 hPa)
flash point   >110 °C
mp   41 - 45 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 40 °C
  Melting range (upper value): ≤ 43 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?