EMAIL THIS PAGE TO A FRIEND
8.05216 EMD Millipore

Suberonitrile

Suberonitrile for synthesis. CAS 629-40-3, EC Number 211-089-2, chemical formula NC(CH₂)₆CN., for synthesis

Synonym: Octanedinitrile, Octanedioic acid dinitrile

  • CAS Number 629-40-3

  • Linear Formula C₈H₁₂N₂

  • Molecular Weight 136.2 g/mol

  •  EC Number 211-089-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   184 - 186 °C (20 hPa)
expl. lim.     8.5 - 29 %(V)
solubility   7 g/l insoluble
density   0.93 g/cm3 (20 °C)
flash point   175 °C
autoignition temp.   455 °C
mp   -3 °C
LD 50 oral   LD50 Rat 242 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.931 - 0.933
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H301: Toxic if swallowed.
Precautionary Statement(s) 
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1 C Combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?