EMAIL THIS PAGE TO A FRIEND
8.05345 EMD Millipore

Lauronitrile

Lauronitrile for synthesis. CAS 2437-25-4, EC Number 219-440-1, chemical formula CH₃(CH₂)₁₀CN., for synthesis

Synonym: Dodecanenitrile, Undecyl cyanide

  • CAS Number 2437-25-4

  • Linear Formula C₁₂H₂₃N

  • Molecular Weight 181.32 g/mol

  •  EC Number 219-440-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8053450100)
bp   197 - 199 °C (133 hPa)
density   0.83 g/cm3 (20 °C)
flash point   113 °C
mp   4 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.823 - 0.826
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3276 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3276 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3276 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?