• USA Home
  • 8.05706 - S-Methylthiouronium sulfate

EMAIL THIS PAGE TO A FRIEND
8.05706 EMD Millipore

S-Methylthiouronium sulfate

S-Methylthiouronium sulfate for synthesis. CAS No. 867-44-7, EC Number 212-759-7., for synthesis

Synonym: S-Methylisothiourea sulfate

  • CAS Number 867-44-7

  • Linear Formula C₄H₁₄N₄O₄S₃

  • Molecular Weight 278.38 g/mol

  •  EC Number 212-759-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Plastic bottle (8057060250)
solubility   260 g/l
density   1.28 g/cm3 (20 °C)
mp   240 - 241 °C (decomposition)
pH   5.5 (260 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 1228 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?