• USA Home
  • 8.05850 - N-Methylcyclohexylamine

EMAIL THIS PAGE TO A FRIEND
8.05850 EMD Millipore

N-Methylcyclohexylamine

N-Methylcyclohexylamine for synthesis. CAS 100-60-7, EC Number 202-869-3, chemical formula C₆H₁₁NHCH₃., for synthesis

Synonym: Methylaminocyclohexane, N-Cyclohexylmethylamine

  • CAS Number 100-60-7

  • Linear Formula C₇H₁₅N

  • Molecular Weight 113.2 g/mol

  •  EC Number 202-869-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8058500100)
bp   148 - 150 °C (1013 hPa)
expl. lim.     2.2 - 10.5 %(V)
vapor pressure   5 hPa (20 °C)
solubility   54 g/l
density   0.86 g/cm3 (20 °C)
flash point   35 °C
autoignition temp.   255 °C
mp   -8 °C
pH   12 (10 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 400 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.856 - 0.859
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 30 99
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H290: May be corrosive to metals.
H302: Harmful if swallowed.
H311: Toxic in contact with skin.
H314: Causes severe skin burns and eye damage.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat.
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2734 , 8 (3), I
Declaration (transport by air) IATA-DGR 
UN 2734 , 8 (3), I
Declaration (transport by sea) IMDG-Code 
UN 2734 , 8 (3), I, Segregation Group: 18 (Alkalis)
Categories of danger 
flammable, harmful, corrosive
R Phrase 
R 10-21/22-34
Flammable.Harmful in contact with skin and if swallowed.Causes burns.
S Phrase 
S 23-26-36/37/39-45
Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?