EMAIL THIS PAGE TO A FRIEND
8.06190 EMD Millipore

Malonamide

Malonamide for synthesis. CAS 108-13-4, EC Number 203-553-8, chemical formula CH₂(CONH₂)₂., for synthesis

Synonym: Malonic acid diamide

  • CAS Number 108-13-4

  • Linear Formula C₃H₆N₂O₂

  • Molecular Weight 102.09 g/mol

  •  EC Number 203-553-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8061900100)
solubility   180 g/l
mp   169 - 172 °C
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 98.0 %
  Melting range (lower value): ≥ 168 °C
  Melting range (upper value): ≤ 173 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?