• USA Home
  • 8.06207 - 3'-Nitroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.06207 EMD Millipore

3'-Nitroacetophenone

3'-Nitroacetophenone for synthesis. CAS No. 121-89-1, EC Number 204-504-3., for synthesis

  • CAS Number 121-89-1

  • Linear Formula C₈H₇NO₃

  • Molecular Weight 165.15 g/mol

  •  EC Number 204-504-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   201 - 203 °C (1013 hPa)
mp   76 - 79 °C
LD 50 oral   LD50 Rat 3250 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 76 °C
  Melting range (upper value): ≤ 79 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?