• USA Home
  • 8.06217 - 4'-Nitroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.06217 EMD Millipore

4'-Nitroacetophenone

4'-Nitroacetophenone for synthesis. CAS 100-19-6, chemical formula 4-(NO₂)C₆H₄COCH₃., for synthesis

Synonym: 1-(4-Nitrophenyl)ethanone

  • CAS Number 100-19-6

  • Linear Formula C₈H₇NO₃

  • Molecular Weight 165.15 g/mol

  •  EC Number 202-827-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8062170005)
flash point   149 °C
mp   76 - 80 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 76 °C
  Melting range (upper value): ≤ 80 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
RTECS 
AM9627000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?