• USA Home
  • 8.06226 - 2-Methoxy-4-nitroaniline

EMAIL THIS PAGE TO A FRIEND
8.06226 EMD Millipore

2-Methoxy-4-nitroaniline

2-Methoxy-4-nitroaniline for synthesis. CAS 97-52-9, chemical formula 2-(CH₃O)-4-(NO₂)C₆H₃NH₂., for synthesis

Synonym: 5-Nitro-2-aminoanisole, 2-Amino-5-nitroanisole, 4-Nitro-o-anisidine

  • CAS Number 97-52-9

  • Linear Formula C₇H₈N₂O₃

  • Molecular Weight 168.15 g/mol

  •  EC Number 202-588-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Glass bottle (8062260250)
solubility   0.2 g/l
autoignition temp.   >360 °C
mp   138 - 141 °C
pH   6.2 (0.2 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 997 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 138 °C
  Melting range (upper value): ≤ 141 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?