EMAIL THIS PAGE TO A FRIEND
8.06291 EMD Millipore

1-Nitronaphthalene

1-Nitronaphthalene for synthesis. CAS 86-57-7, molar mass 173.17 g/mol., for synthesis

  • CAS Number 86-57-7

  • Linear Formula C₁₀H₇NO₂

  • Molecular Weight 173.17 g/mol

  •  EC Number 201-684-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8062910100)
bp   304 °C
vapor pressure   0.00015 hPa (20 °C)
solubility   0.022 g/l
density   1.33 g/cm3 (20 °C)
flash point   170 °C DIN 51758
autoignition temp.   450 °C
mp   59 - 61 °C
LD 50 oral   LD50 Rat 350 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 54 °C
  Melting range (upper value): ≤ 56 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 20 00
Hazard Statement(s) 
H228: Flammable solid.
H302: Harmful if swallowed.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P370 + P378: In case of fire: Use sand for extinction.
Storage class 
4.1B Flammable solid hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2538 , 4.1, III
Declaration (transport by air) IATA-DGR 
UN 2538 , 4.1, III
Declaration (transport by sea) IMDG-Code 
UN 2538 , 4.1, III
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 22-51/53
Harmful if swallowed.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?