• USA Home
  • 8.06294 - 4-Nitro-1,2-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
8.06294 EMD Millipore

4-Nitro-1,2-phenylenediamine

4-Nitro-1,2-phenylenediamine for synthesis. CAS 99-56-9, chemical formula 4-(NO₂)C₆H₃-1,2-(NH₂)₂., for synthesis

Synonym: 1,2-Diamino-4-nitrobenzene

  • CAS Number 99-56-9

  • Linear Formula C₆H₇N₃O₂

  • Molecular Weight 153.14 g/mol

  •  EC Number 202-766-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8062940100)
solubility   1.2 g/l
mp   197 - 202 °C
bulk density   520 kg/m3
LD 50 oral   LD50 Rat 681 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 97.0 %
  Melting range (lower value): ≥ 201 °C
  Melting range (upper value): ≤ 205 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 51 19
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 22-24/25
Do not breathe dust.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?