• USA Home
  • 8.06750 - Ethyl 4-pyridinecarboxylate

EMAIL THIS PAGE TO A FRIEND
8.06750 EMD Millipore

Ethyl 4-pyridinecarboxylate

Ethyl 4-pyridinecarboxylate for synthesis. CAS 1570-45-2, EC Number 216-379-2, chemical formula 4-(C₂H₅OOC)C₅H₄N., for synthesis

Synonym: Isonicotinic acid ethyl ester, Ethyl isonicotinate, 4-Pyridinecarboxylic acid ethyl ester

  • CAS Number 1570-45-2

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 216-379-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8067500100)
bp   220 °C (1013 hPa)
density   1.10 g/cm3 (20 °C)
flash point   93 °C
mp   23 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.102 - 1.105
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
RTECS 
NS1450000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?