EMAIL THIS PAGE TO A FRIEND
8.06805 EMD Millipore

3-Nitrotoluene

3-Nitrotoluene for synthesis. CAS No. 99-08-1, EC Number 202-728-6., for synthesis

Synonym: m-Nitrotoluene

  • CAS Number 99-08-1

  • Linear Formula C₇H₇NO₂

  • Molecular Weight 137.13 g/mol

  •  EC Number 202-728-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8068050100)
expl. lim.     2.2 %(V)
vapor pressure   0.2 hPa (20 °C)
solubility   0.419 g/l
density   1.16 g/cm3 (20 °C)
flash point   98 °C
autoignition temp.   440 °C
mp   16 °C
LD 50 oral   LD50 Rat 1070 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.155 - 1.157
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 20 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H373: May cause damage to organs through prolonged or repeated exposure.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P314: Get medical advice/ attention if you feel unwell.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1664 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 1664 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 1664 , 6.1, II
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 22-33-51/53
Harmful if swallowed.Danger of cumulative effects.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 28-61
After contact with skin, wash immediately with plenty of water.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?