EMAIL THIS PAGE TO A FRIEND
8.06843 EMD Millipore

Nitroethane

Nitroethane for synthesis. CAS 79-24-3, EC Number 201-188-9, chemical formula CH₃CH₂NO₂., for synthesis

  • CAS Number 79-24-3

  • Linear Formula C₂H₅NO₂

  • Molecular Weight 75.07 g/mol

  •  EC Number 201-188-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8068430100)
bp   114 °C (1013 hPa)
expl. lim.     3.4 %(V)
vapor pressure   20 hPa (20 °C)
solubility   45 g/l
density   1.05 g/cm3 (20 °C)
flash point   29 °C
autoignition temp.   410 °C
mp   -90 °C
pH   6 (1 g/l, H₂O, 25 °C)
LD 50 oral   LD50 Rat 1100 mg/kg
LD 50 dermal   LD50 Rabbit > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.049 - 1.051
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 20 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302 + H332: Harmful if swallowed or if inhaled
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2842 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2842 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2842 , 3, III
Categories of danger 
flammable, harmful
R Phrase 
R 10-20/22
Flammable.Harmful by inhalation and if swallowed.
S Phrase 
S 9-25-41
Keep container in a well-ventilated place.Avoid contact with eyes.In case of fire and/or explosion do not breathe fumes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?