EMAIL THIS PAGE TO A FRIEND
8.06903 EMD Millipore

1-Chlorooctane

1-Chlorooctane for synthesis. CAS 111-85-3, chemical formula CH₃(CH₂)₇Cl., for synthesis

Synonym: Octyl chloride

  • CAS Number 111-85-3

  • Linear Formula C₈H₁₇Cl

  • Molecular Weight 148.67 g/mol

  •  EC Number 203-915-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8069030100)
bp   181 - 183 °C (1013 hPa)
vapor pressure   1.5 hPa (20 °C)
solubility   0.02 g/l
density   0.871 - 0.875 g/cm3 (20 °C)
flash point   68 °C
autoignition temp.   230 °C
mp   -58 °C
LD 50 oral   LD50 Rat > 20000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.871 - 0.875
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 19 00
Hazard Statement(s) 
H304: May be fatal if swallowed and enters airways.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
Categories of danger 
dangerous for the environment
R Phrase 
R 50/53
Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?