EMAIL THIS PAGE TO A FRIEND
8.06921 EMD Millipore

1,8-Octanediol

1,8-Octanediol for synthesis. CAS 629-41-4, molar mass 146.23 g/mol., for synthesis

Synonym: Octamethylene glycol

  • CAS Number 629-41-4

  • Linear Formula C₈H₁₈O₂

  • Molecular Weight 146.23 g/mol

  •  EC Number 211-090-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8069210025)
bp   171 - 173 °C (27 hPa)
flash point   175 °C
autoignition temp.   270 °C
mp   58 - 61 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 57 °C
  Melting range (upper value): ≤ 61 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 39 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?