• USA Home
  • 8.06927 - 2,2'-Oxydiethanethiol

EMAIL THIS PAGE TO A FRIEND
8.06927 EMD Millipore

2,2'-Oxydiethanethiol

for synthesis

Synonym: Bis(2-mercaptoethyl) ether

  • CAS Number 2150-02-9

  • Linear Formula C₄H₁₀OS₂

  • Molecular Weight 138.24 g/mol

  •  EC Number 218-421-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass Bottle in Metal can (8069270025)
bp   216 - 218 °C
density   1.12 g/cm3 (20 °C)
flash point   98 °C
mp   -80 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.114 - 1.117
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?